Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Diiodo-L-tyrosine, 95%

Click to view available options
Quantity:
25g
100g
300-39-0
C9H9I2NO3
432.984
MFCD00016542
NYPYHUZRZVSYKL-ZETCQYMHSA-N
3,5-diiodo-l-tyrosine,l-3,5-diiodotyrosine,diiodotyrosine,l-diiodotyrosine,3,5-diiodotyrosine,3,5-l-diiodotyrosine,3,5-iodo-l-tyrosine,h-tyr 3,5-i2-oh,dit amino acid,l-tyrosine, 3,5-diiodo
9305
CHEBI:15768
(2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid
C1=C(C=C(C(=C1I)O)I)CC(C(=O)O)N
300-39-0
C9H9I2NO3
432.984
MFCD00016542
NYPYHUZRZVSYKL-ZETCQYMHSA-N
3,5-diiodo-l-tyrosine,l-3,5-diiodotyrosine,diiodotyrosine,l-diiodotyrosine,3,5-diiodotyrosine,3,5-l-diiodotyrosine,3,5-iodo-l-tyrosine,h-tyr 3,5-i2-oh,dit amino acid,l-tyrosine, 3,5-diiodo
9305
CHEBI:15768
(2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid
C1=C(C=C(C(=C1I)O)I)CC(C(=O)O)N
Beilstein 2218691
Formula Weight 432.98
Physical Form Solid
Percent Purity 95%
Color Yellow
Quantity 100g
Sensitivity Light Sensitive
Chemical Name or Material 3,5-Diiodo-L-tyrosine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA