Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diiodo-L-tyrosine, Dihydrate, Approx. 99%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
18835-59-1
C9H13I2NO5
469.01
YWAGQOOMOOUEGY-UHFFFAOYNA-N
2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid dihydrate
O.O.NC(CC1=CC(I)=C(O)C(I)=C1)C(O)=O
18835-59-1
C9H13I2NO5
469.01
YWAGQOOMOOUEGY-UHFFFAOYNA-N
2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid dihydrate
O.O.NC(CC1=CC(I)=C(O)C(I)=C1)C(O)=O

D2348, 18835-59-1, C9H9I2NO3•2H2O

Spectrum™ 3,5-Diiodo-L-tyrosine, Dihydrate , also known as Diiodotyrosine or DIT, is a precursor used in producing thyroid hormone; it is also a modulator of the enzyme thyroid peroxidase. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 25 g
CAS Max % 100%
Percent Purity Approx. 99%