Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2478-38-8
C10H12O4
196.2
MFCD00008748
OJOBTAOGJIWAGB-UHFFFAOYSA-N
acetosyringone,3',5'-dimethoxy-4'-hydroxyacetophenone,1-4-hydroxy-3,5-dimethoxyphenyl ethanone,4'-hydroxy-3',5'-dimethoxyacetophenone,3,5-dimethoxy-4-hydroxyacetophenone,acetosyringon,ethanone, 1-4-hydroxy-3,5-dimethoxyphenyl,1-4-hydroxy-3,5-dimethoxyphenyl ethan-1-one,acetosyringenin,acetophenone, 4'-hydroxy-3',5'-dimethoxy
17198
CHEBI:2404
1-(4-hydroxy-3,5-dimethoxyphenyl)ethanone
CC(=O)C1=CC(=C(C(=C1)OC)O)OC
2478-38-8
C10H12O4
196.2
MFCD00008748
OJOBTAOGJIWAGB-UHFFFAOYSA-N
acetosyringone,3',5'-dimethoxy-4'-hydroxyacetophenone,1-4-hydroxy-3,5-dimethoxyphenyl ethanone,4'-hydroxy-3',5'-dimethoxyacetophenone,3,5-dimethoxy-4-hydroxyacetophenone,acetosyringon,ethanone, 1-4-hydroxy-3,5-dimethoxyphenyl,1-4-hydroxy-3,5-dimethoxyphenyl ethan-1-one,acetosyringenin,acetophenone, 4'-hydroxy-3',5'-dimethoxy
17198
CHEBI:2404
1-(4-hydroxy-3,5-dimethoxyphenyl)ethanone
CC(=O)C1=CC(=C(C(=C1)OC)O)OC
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 123°C to 128°C
Color Beige to Brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula HOC6H2(OCH3)2COCH3
Beilstein 08,II,444
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,acetone,ether
Formula Weight 196.2
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 3′,5′-Dimethoxy-4′-hydroxyacetophenone, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 219-610-5

RTECSNumber : AM8440000

TSCA : TSCA