Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethoxy-4-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number AC132880050
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
134-96-3
C9H10O4
182.18
MFCD00006943
KCDXJAYRVLXPFO-UHFFFAOYSA-N
syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde
8655
CHEBI:67380
4-hydroxy-3,5-dimethoxybenzaldehyde
COC1=CC(C=O)=CC(OC)=C1O
134-96-3
C9H10O4
182.18
MFCD00006943
KCDXJAYRVLXPFO-UHFFFAOYSA-N
syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde
8655
CHEBI:67380
4-hydroxy-3,5-dimethoxybenzaldehyde
COC1=CC(C=O)=CC(OC)=C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 110.0°C to 113.0°C
Color Brown to Green-Yellow
Density 1.0130g/mL
Boiling Point 192.0°C to 193.0°C (14.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.013
Merck Index 15, 9145
Solubility Information Solubility in water: insoluble. Other solubilities: very sparingly soluble in petr ether, soluble in alcohol, ether, chloroform, hot benzene, glacial acetic acid
Formula Weight 182.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxy-4-hydroxybenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 205-167-5

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit