Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
530-59-6
C11H12O5
224.21
PCMORTLOPMLEFB-ONEGZZNKSA-N
sinapic acid,sinapinic acid,3,5-dimethoxy-4-hydroxycinnamic acid,sinapinate,sinapate,trans-sinapic acid,4-hydroxy-3,5-dimethoxycinnamic acid,synapoic acid,2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid,unii-p0i60993ec
637775
CHEBI:15714
(E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
COC1=CC(=CC(=C1O)OC)C=CC(=O)O
530-59-6
C11H12O5
224.21
PCMORTLOPMLEFB-ONEGZZNKSA-N
sinapic acid,sinapinic acid,3,5-dimethoxy-4-hydroxycinnamic acid,sinapinate,sinapate,trans-sinapic acid,4-hydroxy-3,5-dimethoxycinnamic acid,synapoic acid,2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid,unii-p0i60993ec
637775
CHEBI:15714
(E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
COC1=CC(=CC(=C1O)OC)C=CC(=O)O
Packaging Glass bottle
Melting Point 203.0°C to 205.0°C
Infrared Spectrum Authentic
Quantity 1g
Color Beige-Yellow to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HOC6H2(OCH3)2CH=CHCO2H
Beilstein 10,508
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in methanol,ethanol and acetone
Formula Weight 224.21
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 3, 5-Dimethoxy-4-hydroxycinnamic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.