Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3',5'-Dimethoxyacetophenone, 97%

Click to view available options
Quantity:
1g
5g
25g
39151-19-4
C10H12O3
180.203
MFCD00008739
YJKHOUIVWKQRSL-UHFFFAOYSA-N
3',5'-dimethoxyacetophenone,1-3,5-dimethoxyphenyl ethanone,3,5-dimethoxyacetophenone,1-3,5-dimethoxyphenyl ethan-1-one,ethanone, 1-3,5-dimethoxyphenyl,1-acetyl-3,5-dimethoxybenzene,acetophenone, 3',5'-dimethoxy-6ci,7ci,acmc-20anci,pubchem3391,opera_id_1889
95997
1-(3,5-dimethoxyphenyl)ethanone
CC(=O)C1=CC(=CC(=C1)OC)OC
39151-19-4
C10H12O3
180.203
MFCD00008739
YJKHOUIVWKQRSL-UHFFFAOYSA-N
3',5'-dimethoxyacetophenone,1-3,5-dimethoxyphenyl ethanone,3,5-dimethoxyacetophenone,1-3,5-dimethoxyphenyl ethan-1-one,ethanone, 1-3,5-dimethoxyphenyl,1-acetyl-3,5-dimethoxybenzene,acetophenone, 3',5'-dimethoxy-6ci,7ci,acmc-20anci,pubchem3391,opera_id_1889
95997
1-(3,5-dimethoxyphenyl)ethanone
CC(=O)C1=CC(=CC(=C1)OC)OC
Applications
3′,5′-Dimethoxyacetophenone is used as starting reagent in the synthesis of 5,7-dimethoxy-3-methylindazole1. It was used in the synthesis of (R)-(+)-1-(3,5-dimethoxyphenyl)ethyl acetate-ether-180.

Solubility
Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Refractive Index 1.543
Boiling Point 290°C to 291°C
Quantity 5g
Flash Point >110°C (230°F)
Melting Point 39°C to 41°C
Beilstein 2045586
Solubility Information Insoluble in water.
Formula Weight 180.2
Percent Purity 97%
Chemical Name or Material 3',5'-Dimethoxyacetophenone

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 254-322-3

RTECSNumber : KM5775717

TSCA : No

Recommended Storage : Ambient temperatures