Alfa Aesar™ 3',5'-Dimethoxyacetophenone, 97%

Manufacturer: Alfa Aesar™ B2522414

Quantity 25g

 View more versions of this product

Catalog No. AAB2522414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 39151-19-4
    Molecular Formula C10H12O3
    Formula Weight 180.2
    MDL Number MFCD00008739
    Beilstein 2045586
    Chemical Name or Material 3',5'-Dimethoxyacetophenone

    Specifications

    Formula Weight 180.2
    Molecular Formula C10H12O3
    Melting Point 39° to 41°C
    Boiling Point 290° to 291°C
    MDL Number MFCD00008739
    CAS 39151-19-4
    Beilstein 2045586
    Quantity 25g
    Flash Point >110°C (230°F)
    RTECS Number KM5775717
    Chemical Name or Material 3',5'-Dimethoxyacetophenone
    Assay 97%

    Description

    Applications
    3 ft.,5 ft.-Dimethoxyacetophenone is used as starting reagent in the synthesis of 5,7-dimethoxy-3-methylindazole1. It was used in the synthesis of (R)-(+)-1-(3,5-dimethoxyphenyl)ethyl acetate-ether-180.

    Solubility
    Insoluble in water.

    Notes
    Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit