Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethoxyaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
10272-07-8
C8H11NO2
153.18
MFCD00008392
WNRGWPVJGDABME-UHFFFAOYSA-N
benzenamine, 3,5-dimethoxy,3,5-dimethoxybenzeneamine,5-aminoresorcinol dimethyl ether,3,5-dimethoxy-phenylamine,3,5-dimethoxyanaline,3,5-dimethoxy aniline,5-aminoresorcinol dimethylether,3,5-dimethoxybenzenamine,3,5-dimethoxyphenylamine,pubchem4423
66301
3,5-dimethoxyaniline
COC1=CC(=CC(=C1)N)OC
10272-07-8
C8H11NO2
153.18
MFCD00008392
WNRGWPVJGDABME-UHFFFAOYSA-N
benzenamine, 3,5-dimethoxy,3,5-dimethoxybenzeneamine,5-aminoresorcinol dimethyl ether,3,5-dimethoxy-phenylamine,3,5-dimethoxyanaline,3,5-dimethoxy aniline,5-aminoresorcinol dimethylether,3,5-dimethoxybenzenamine,3,5-dimethoxyphenylamine,pubchem4423
66301
3,5-dimethoxyaniline
COC1=CC(=CC(=C1)N)OC
Boiling Point 178.0°C (20.0 mmHg)
Infrared Spectrum Authentic
Color Brown to Colorless
Melting Point 53.0°C to 55.0°C
Flash Point 135°C
Quantity 5g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3O)2C6H3NH2
Solubility Information Solubility in water: insoluble
Formula Weight 153.18
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3, 5-Dimethoxyaniline, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 233-616-5

TSCA : TSCA