Alfa Aesar™ 3,5-Dimethoxybenzamide, 98%

Manufacturer: Alfa Aesar™ B2481706

Quantity 5g

 View more versions of this product

Catalog No. AAB2481706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17213-58-0
    Molecular Formula C9H11NO3
    Formula Weight 181.19
    MDL Number MFCD00007987
    Beilstein 2641361
    Chemical Name or Material 3,5-Dimethoxybenzamide

    Specifications

    Formula Weight 181.19
    Molecular Formula C9H11NO3
    Melting Point 142° to 146°C
    MDL Number MFCD00007987
    Beilstein 2641361
    Quantity 5g
    CAS 17213-58-0
    Chemical Name or Material 3,5-Dimethoxybenzamide
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit