Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dimethoxybenzonitrile, 98%

Click to view available options
Quantity:
5g
25g
100g
19179-31-8
C9H9NO2
163.176
MFCD00001804
NVTHWSJNXVDIKR-UHFFFAOYSA-N
benzonitrile, 3,5-dimethoxy,3,5-dimethoxy benzonitrile,3,5-dimethoxybenzenecarbonitrile,pubchem3122,benzonitrile,5-dimethoxy,acmc-209z2h,3,5-dimethoxybenzo nitrile,3-10-00-01449 beilstein handbook reference,ksc497i3p,1-cyano-3,5-dimethoxybenzene
29482
3,5-dimethoxybenzonitrile
COC1=CC(=CC(=C1)C#N)OC
19179-31-8
C9H9NO2
163.176
MFCD00001804
NVTHWSJNXVDIKR-UHFFFAOYSA-N
benzonitrile, 3,5-dimethoxy,3,5-dimethoxy benzonitrile,3,5-dimethoxybenzenecarbonitrile,pubchem3122,benzonitrile,5-dimethoxy,acmc-209z2h,3,5-dimethoxybenzo nitrile,3-10-00-01449 beilstein handbook reference,ksc497i3p,1-cyano-3,5-dimethoxybenzene
29482
3,5-dimethoxybenzonitrile
COC1=CC(=CC(=C1)C#N)OC
Quantity 5g
Melting Point 87°C to 89°C
UN Number UN3439
Formula Weight 163.18
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxybenzonitrile

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.