Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dimethoxybenzyl alcohol, 99%

Manufacturer:  Alfa Aesar™ B2513803

Quantity 1g

 View more versions of this product

Catalog No. AAB2513803


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

705-76-0
168.19
1866305
C9H12O3
MFCD00004641
3,5-Dimethoxybenzyl alcohol

Specifications

705-76-0
46° to 50°C
168.19
1866305
>110°C (230°F)
99%
C9H12O3
183° to 186°C (20mmHg)
MFCD00004641
1g
3,5-Dimethoxybenzyl alcohol
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit