Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethoxycinnamic Acid, Predominantly trans Isomer 99+%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
16909-11-8
C11H12O4
208.213
MFCD00004389
VLSRUFWCGBMYDJ-ONEGZZNKSA-N
3,5-dimethoxycinnamic acid,3-3,5-dimethoxyphenyl acrylic acid,e-3-3,5-dimethoxyphenyl acrylic acid,2e-3-3,5-dimethoxyphenyl prop-2-enoic acid,3-3,5-dimethoxyphenyl-2-propenoic acid,trans-3,5-dimethoxycinnamic acid,3-3,5-dimethoxyphenyl prop-2-enoic acid,pubchem8295,3,5-dimethoxycinnamicacid,rarechem bk hw 0037
5324677
(E)-3-(3,5-dimethoxyphenyl)prop-2-enoic acid
COC1=CC(=CC(=C1)C=CC(=O)O)OC
16909-11-8
C11H12O4
208.213
MFCD00004389
VLSRUFWCGBMYDJ-ONEGZZNKSA-N
3,5-dimethoxycinnamic acid,3-3,5-dimethoxyphenyl acrylic acid,e-3-3,5-dimethoxyphenyl acrylic acid,2e-3-3,5-dimethoxyphenyl prop-2-enoic acid,3-3,5-dimethoxyphenyl-2-propenoic acid,trans-3,5-dimethoxycinnamic acid,3-3,5-dimethoxyphenyl prop-2-enoic acid,pubchem8295,3,5-dimethoxycinnamicacid,rarechem bk hw 0037
5324677
(E)-3-(3,5-dimethoxyphenyl)prop-2-enoic acid
COC1=CC(=CC(=C1)C=CC(=O)O)OC
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 10g
Melting Point 173°C to 176°C
Color Beige-Yellow or Green to White
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula (CH3O)2C6H3CH=CHCO2H
Beilstein 10, II, 297
Formula Weight 208.21
Physical Form Crystalline Powder or Needles
Percent Purity ≥99.0%
Chemical Name or Material 3, 5-Dimethoxycinnamic acid, 99+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 240-956-8

TSCA : TSCA