Alfa Aesar™ 3,5-Dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole-4-boronic acid neopentyl glycol ester, 95%

Manufacturer: Alfa Aesar™ H3272603

Quantity 1g

 View more versions of this product

Catalog No. AAH3272603

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C15H25BN2O3
    Formula Weight 292.19
    MDL Number MFCD09837628
    Chemical Name or Material 3,5-Dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole-4-boronic acid neopentyl glycol ester

    Specifications

    Formula Weight 292.19
    Molecular Formula C15H25BN2O3
    MDL Number MFCD09837628
    Quantity 1g
    Chemical Name or Material 3,5-Dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole-4-boronic acid neopentyl glycol ester
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit