Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dimethyl-1-adamantaneacetic acid, 97%

Click to view available options
Quantity:
1g
5g
14202-14-3
C14H22O2
222.328
MFCD00077207
FUOXJVUIQUYDDI-UHFFFAOYSA-N
3,5-dimethyl-1-adamantyl acetic acid,3,5-dimethyl-1-adamantaneacetic acid,3,5-dimethyladamantane-1-acetic acid,2-3,5-dimethyl-1-adamantyl acetic acid,2-3,5-dimethyladamantan-1-yl acetic acid,3,5-dimethyladamantan-1-yl acetic acid,2-3,5-dimethyladamantanyl acetic acid,3,5-dimethyltricyclo 3.3.1.1~3,7~ dec-1-yl acetic acid,acmc-20ancc,3,5-dimethyladamantaneacetic acid
4067589
2-(3,5-dimethyl-1-adamantyl)acetic acid
CC12CC3CC(C1)(CC(C3)(C2)CC(=O)O)C
14202-14-3
C14H22O2
222.328
MFCD00077207
FUOXJVUIQUYDDI-UHFFFAOYSA-N
3,5-dimethyl-1-adamantyl acetic acid,3,5-dimethyl-1-adamantaneacetic acid,3,5-dimethyladamantane-1-acetic acid,2-3,5-dimethyl-1-adamantyl acetic acid,2-3,5-dimethyladamantan-1-yl acetic acid,3,5-dimethyladamantan-1-yl acetic acid,2-3,5-dimethyladamantanyl acetic acid,3,5-dimethyltricyclo 3.3.1.1~3,7~ dec-1-yl acetic acid,acmc-20ancc,3,5-dimethyladamantaneacetic acid
4067589
2-(3,5-dimethyl-1-adamantyl)acetic acid
CC12CC3CC(C1)(CC(C3)(C2)CC(=O)O)C
Applications
A commonly used as an organic reagent and pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Melting Point 110°C to 115°C
Quantity 1g
Color White
Formula Weight 222.32
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 3,5-Dimethyl-1-adamantaneacetic acid