Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dimethyl-1-phenyl-1H-pyrazole, 97%

Click to view available options
Quantity:
5g
25g
1131-16-4
C11H12N2
172.231
MFCD00020728
ULPMPUPEFBDQQA-UHFFFAOYSA-N
3,5-dimethyl-1-phenyl-1h-pyrazole,1h-pyrazole, 3,5-dimethyl-1-phenyl,dimethylphenylpyrazole,unii-0sb52bxx7m,1h-pyrazole,3,5-dimethyl-1-phenyl,0sb52bxx7m,1,3-dimethyl-5-phenylpyrazole,phenyldimethylpyrazole,enamine_001158,pyrazole, dimethylphenyl
70800
3,5-dimethyl-1-phenylpyrazole
CC1=CC(=NN1C2=CC=CC=C2)C
1131-16-4
C11H12N2
172.231
MFCD00020728
ULPMPUPEFBDQQA-UHFFFAOYSA-N
3,5-dimethyl-1-phenyl-1h-pyrazole,1h-pyrazole, 3,5-dimethyl-1-phenyl,dimethylphenylpyrazole,unii-0sb52bxx7m,1h-pyrazole,3,5-dimethyl-1-phenyl,0sb52bxx7m,1,3-dimethyl-5-phenylpyrazole,phenyldimethylpyrazole,enamine_001158,pyrazole, dimethylphenyl
70800
3,5-dimethyl-1-phenylpyrazole
CC1=CC(=NN1C2=CC=CC=C2)C
Quantity 25g
Flash Point >110°C (230°F)
Boiling Point 144°C to 145°C (12mmHg)
Beilstein 132902
Formula Weight 172.23
Percent Purity 97%
Chemical Name or Material 3,5-Dimethyl-1-phenyl-1H-pyrazole