Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-1H-pyrazole-1-acetic acid, 95%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
16034-49-4
C7H10N2O2
154.17
JYSWEDYPQJOEPO-UHFFFAOYSA-N
2-3,5-dimethyl-1h-pyrazol-1-yl acetic acid,3,5-dimethyl-pyrazol-1-yl-acetic acid,3,5-dimethyl-1h-pyrazol-1-yl acetic acid,3,5-dimethylpyrazol-1-yl acetic acid,1h-pyrazole-1-acetic acid, 3,5-dimethyl,3-nitro-benzene-thiol,3,5-dimethyl-1h-pyrazole-1-acetic acid,1h-pyrazole-1-aceticacid, 3,5-dimethyl,2-3,5-dimethylpyrazolyl acetic acid,3,5-dimethyl-pyrazol-1-yl acetic acid
572875
2-(3,5-dimethylpyrazol-1-yl)acetic acid
CC1=CC(=NN1CC(=O)O)C
16034-49-4
C7H10N2O2
154.17
JYSWEDYPQJOEPO-UHFFFAOYSA-N
2-3,5-dimethyl-1h-pyrazol-1-yl acetic acid,3,5-dimethyl-pyrazol-1-yl-acetic acid,3,5-dimethyl-1h-pyrazol-1-yl acetic acid,3,5-dimethylpyrazol-1-yl acetic acid,1h-pyrazole-1-acetic acid, 3,5-dimethyl,3-nitro-benzene-thiol,3,5-dimethyl-1h-pyrazole-1-acetic acid,1h-pyrazole-1-aceticacid, 3,5-dimethyl,2-3,5-dimethylpyrazolyl acetic acid,3,5-dimethyl-pyrazol-1-yl acetic acid
572875
2-(3,5-dimethylpyrazol-1-yl)acetic acid
CC1=CC(=NN1CC(=O)O)C
Packaging Glass Bottle
Melting Point 186.0°C to 188.0°C
Infrared Spectrum Authentic
Quantity 1g
Assay Percent Range 94% min. (HPLC)
Formula Weight 154.17
Percent Purity 95%
Chemical Name or Material 3,5-Dimethyl-1H-pyrazole-1-acetic acid

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Warning