Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-4-hydroxybenzonitrile, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
4198-90-7
C9H9NO
147.18
MFCD00221716
WFYGXOWFEIOHCZ-UHFFFAOYSA-N
3,5-dimethyl-4-hydroxybenzonitrile,4-cyano-2,6-dimethylphenol,2,6-dimethyl-4-cyanophenol,4-hydroxy-3,5-dimethyl-benzonitrile,3,5-dimethyl-4-hydroxy benzonitrile,benzonitrile, 3,5-dimethyl-4-hydroxy,4-hydroxy-3,5-dimethylbenzenecarbonitrile,benzonitrile, 4-hydroxy-3,5-dimethyl,pubchem11065,acmc-209jmm
20176
4-hydroxy-3,5-dimethylbenzonitrile
CC1=CC(=CC(=C1O)C)C#N
4198-90-7
C9H9NO
147.18
MFCD00221716
WFYGXOWFEIOHCZ-UHFFFAOYSA-N
3,5-dimethyl-4-hydroxybenzonitrile,4-cyano-2,6-dimethylphenol,2,6-dimethyl-4-cyanophenol,4-hydroxy-3,5-dimethyl-benzonitrile,3,5-dimethyl-4-hydroxy benzonitrile,benzonitrile, 3,5-dimethyl-4-hydroxy,4-hydroxy-3,5-dimethylbenzenecarbonitrile,benzonitrile, 4-hydroxy-3,5-dimethyl,pubchem11065,acmc-209jmm
20176
4-hydroxy-3,5-dimethylbenzonitrile
CC1=CC(=CC(=C1O)C)C#N
Melting Point 123.0°C to 126.0°C
Color White to Beige
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 10, IV, 681
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol,chloroform
Formula Weight 147.18
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethyl-4-hydroxybenzonitrile
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

RTECSNumber : DI4359000

TSCA : TSCA