Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-4-methoxyphenylboronic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
301699-39-8
C9H13BO3
180.01
WZUCSPWZHRVOSD-UHFFFAOYSA-N
3,5-dimethyl-4-methoxyphenylboronic acid,3,5-dimethyl-4-methoxybenzeneboronic acid,4-methoxy-3,5-dimethylphenyl boronic acid,4-methoxy-3,5-dimethyl-phenyl boronic acid,4-methoxy-3,5-dimethylbenzeneboronic acid,4-methoxy-3,5-dimethylphenyl boranediol,boronic acid, 4-methoxy-3,5-dimethylphenyl,pubchem6937,acmc-209hdl,3,5-dimethyl-4-methoxybenzeneboronicacid
3817586
301699-39-8
C9H13BO3
180.01
WZUCSPWZHRVOSD-UHFFFAOYSA-N
3,5-dimethyl-4-methoxyphenylboronic acid,3,5-dimethyl-4-methoxybenzeneboronic acid,4-methoxy-3,5-dimethylphenyl boronic acid,4-methoxy-3,5-dimethyl-phenyl boronic acid,4-methoxy-3,5-dimethylbenzeneboronic acid,4-methoxy-3,5-dimethylphenyl boranediol,boronic acid, 4-methoxy-3,5-dimethylphenyl,pubchem6937,acmc-209hdl,3,5-dimethyl-4-methoxybenzeneboronicacid
3817586
Melting Point 203°C to 204°C
Quantity 10g
Assay Percent Range 94% min. (HPLC)
Formula Weight 180.01
Percent Purity 95%
Chemical Name or Material 3, 5-Dimethyl-4-methoxyphenylboronic acid

GHS Signal Word: Warning