Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dimethyl-4-methoxyphenylmagnesium bromide, 0.5M in 2-MeTHF

Manufacturer:  Alfa Aesar™ H54781AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH54781AE


  Check Availability
Add to cart

Specifications

Description

Applications
3,5-Dimethyl-4-methoxyphenylmagnesium bromide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to air and moisture. Protect from humidity and water. Store under dry inert gas.
Description & Specifications

Chemical Identifiers

185416-17-5
239.39
UN2924
C9H11BrMgO
MFCD13319917
3,5-Dimethyl-4-methoxyphenylmagnesium bromide

Specifications

185416-17-5
239.39
MFCD13319917
100mL
C9H11BrMgO
0.5M in 2-MeTHF
UN2924
3,5-Dimethyl-4-methoxyphenylmagnesium bromide

Safety and Handling

GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit