Alfa Aesar™ 3,5-Dimethyladamantane-1-methanol, 97%

Manufacturer: Alfa Aesar™ H3784506

Quantity 5g

 View more versions of this product

Catalog No. AAH3784506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 26919-42-6
    Formula Weight 194.32
    MDL Number MFCD00188062
    Synonym 3,5-Dimethyl(1-hydroxymethyl)adamantane
    Chemical Name or Material 3,5-Dimethyladamantane-1-methanol

    Specifications

    Melting Point 55° to 56°C
    Formula Weight 194.32
    Boiling Point 114°C (2.25mmHg)
    Quantity 5g
    Physical Form Solid
    MDL Number MFCD00188062
    Synonym 3,5-Dimethyl(1-hydroxymethyl)adamantane
    Color White
    CAS 26919-42-6
    Chemical Name or Material 3,5-Dimethyladamantane-1-methanol
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit