Learn More
3,5-Dimethylaniline, 97+%, Thermo Scientific Chemicals
Description
3,5-Dimethylaniline is the reagent commonly useful in the manufacture of dyes. It undergoes polymerization in the presence of cerium(IV) sulfate as an oxidant. 3,5-Dimethylaniline was used in the synthesis of chiral packing materials for high performance liquid chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3,5-Dimethylaniline is the reagent commonly useful in the manufacture of dyes. It undergoes polymerization in the presence of cerium(IV) sulfate as an oxidant. 3,5-Dimethylaniline was used in the synthesis of chiral packing materials for high performance liquid chromatography.
Solubility
Soluble in water. <0.1 g/100 mL at 19°C
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Air and light sensitive. Incompatible to acids, acid chlorides, acid anhydrides, oxidizing agents, chloroformates, halogens.
Specifications
Melting Point | 9°C to 11°C |
Density | 0.971 |
Boiling Point | 220°C to 221°C |
Quantity | 500 g |
Odor | Characteristic |
Refractive Index | 1.558 |
UN Number | UN1711 |
Beilstein | 507051 |
Merck Index | 14,10084 |
Solubility Information | Soluble in water. <0.1g/100 mL at 19°C |
Show More |
Safety and Handling
Hazard Category | H227-H301+H311+H331-H373 |
Hazard Statement | GHS H Statement H301-H311-H331-H373-H227 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Combustible liquid. |
Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLIDINES, LIQUID |
EINECSNumber | 203-607-0 |
RTECSNumber | ZE9625000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.