Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylanisole, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
874-63-5
C9H12O
136.19
MFCD00008398
JCHJBEZBHANKGA-UHFFFAOYSA-N
3,5-dimethylanisole,5-methoxy-m-xylene,benzene, 1-methoxy-3,5-dimethyl,1-methoxy-3,5-dimethyl-benzene,3,5-dimethylanisol,3,5-dimethylanizole,acmc-1bkxt,anisole, 3,5-dimethyl,ksc494e7h,5-methoxy-1,3-dimethylbenzene
70126
1-methoxy-3,5-dimethylbenzene
CC1=CC(=CC(=C1)OC)C
874-63-5
C9H12O
136.19
MFCD00008398
JCHJBEZBHANKGA-UHFFFAOYSA-N
3,5-dimethylanisole,5-methoxy-m-xylene,benzene, 1-methoxy-3,5-dimethyl,1-methoxy-3,5-dimethyl-benzene,3,5-dimethylanisol,3,5-dimethylanizole,acmc-1bkxt,anisole, 3,5-dimethyl,ksc494e7h,5-methoxy-1,3-dimethylbenzene
70126
1-methoxy-3,5-dimethylbenzene
CC1=CC(=CC(=C1)OC)C
Boiling Point 193°C
Flash Point 65°C
Specific Gravity 0.96
Density 0.9600g/mL
Refractive Index 1.5117 to 1.5137
Color Colorless to Yellow
Quantity 100g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2C6H3OCH3
Beilstein 06,493
Formula Weight 136.19
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 3, 5-Dimethylanisole
Show More Show Less