Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylbenzaldehyde 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
5779-95-3
C9H10O
134.178
MFCD00082777
NBEFMISJJNGCIZ-UHFFFAOYSA-N
benzaldehyde, 3,5-dimethyl,3,5-dimethyl benzaldehyde,m-xylene-5-carboxaldehyde,3,5-dimethyl-benzaldehyde,pubchem15515,acmc-1arkp,3,5-dimehtylbenzaldehyde,#,ksc494c0b,3,5-dimethylbenzaldehyde
34225
3,5-dimethylbenzaldehyde
CC1=CC(=CC(=C1)C=O)C
5779-95-3
C9H10O
134.178
MFCD00082777
NBEFMISJJNGCIZ-UHFFFAOYSA-N
benzaldehyde, 3,5-dimethyl,3,5-dimethyl benzaldehyde,m-xylene-5-carboxaldehyde,3,5-dimethyl-benzaldehyde,pubchem15515,acmc-1arkp,3,5-dimehtylbenzaldehyde,#,ksc494c0b,3,5-dimethylbenzaldehyde
34225
3,5-dimethylbenzaldehyde
CC1=CC(=CC(=C1)C=O)C
Quantity 5g
Color Colorless to Yellow
Packaging Glass bottle
Density 0.9980g/mL
Infrared Spectrum Authentic
Refractive Index 1.5370 to 1.5410 (20°C, 589nm)
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Linear Formula (CH3)2C6H3COH
Formula Weight 134.18
Physical Form Liquid
Percent Purity ≥97.0%
Chemical Name or Material 3, 5-Dimethylbenzaldehyde
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.