Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dimethylbenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
172975-69-8
C8H11BO2
149.984
MFCD00185689
DJGHSJBYKIQHIK-UHFFFAOYSA-N
3,5-dimethylphenyl boronic acid,3,5-dimethylbenzeneboronic acid,3,5-dimethylphenyl boranediol,m-xylene-5-boronic acid,boronic acid, 3,5-dimethylphenyl,pubchem1833,acmc-209e5l,ksc174i6l,3.5-dimethylphenylboronic acid
2734349
(3,5-dimethylphenyl)boronic acid
B(C1=CC(=CC(=C1)C)C)(O)O
172975-69-8
C8H11BO2
149.984
MFCD00185689
DJGHSJBYKIQHIK-UHFFFAOYSA-N
3,5-dimethylphenyl boronic acid,3,5-dimethylbenzeneboronic acid,3,5-dimethylphenyl boranediol,m-xylene-5-boronic acid,boronic acid, 3,5-dimethylphenyl,pubchem1833,acmc-209e5l,ksc174i6l,3.5-dimethylphenylboronic acid
2734349
(3,5-dimethylphenyl)boronic acid
B(C1=CC(=CC(=C1)C)C)(O)O
Applications
3,5-Dimethylbenzeneboronic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Quantity 1g
Melting Point 235°C to 237°C
Beilstein 7368658
Formula Weight 149.99
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning