Alfa Aesar™ 3,5-Dimethylbenzoic acid, 98+%

Manufacturer: Alfa Aesar™ A1119818

Quantity 50g

 View more versions of this product

Catalog No. AAA1119818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 499-06-9
    Molecular Formula C9H10O2
    Formula Weight 150.18
    MDL Number MFCD00002525
    Beilstein 1072182
    Synonym Mesitylenic acid
    Chemical Name or Material 3,5-Dimethylbenzoic acid

    Specifications

    Formula Weight 150.18
    MDL Number MFCD00002525
    Quantity 50g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Molecular Formula C9H10O2
    CAS 499-06-9
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Melting Point 168° to 172°C
    Beilstein 1072182
    Odor Odorless
    Synonym Mesitylenic acid
    RTECS Number DG8734030
    Chemical Name or Material 3,5-Dimethylbenzoic acid
    Assay ≥98%

    Description

    Applications
    3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp

    Solubility
    Soluble in methanol. (1 g/10 mL). Slightly soluble in water.

    Notes
    Store away from oxidizing agents. Use only in a chemical fume hood. Incompatible with strong acids and bases.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit