Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylbenzyl bromide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
27129-86-8
C9H11Br
199.09
MFCD00013539
QXDHXCVJGBTQMK-UHFFFAOYSA-N
3,5-dimethylbenzyl bromide,1-bromomethyl-3,5-dimethylbenzene,3,5-dimethylbenzylbromide,alpha-bromomesitylene,benzene, 1-bromomethyl-3,5-dimethyl,.alpha.-bromomesitylene,5-bromomethyl-1,3-dimethylbenzene,a-bromomesitylene,pubchem9979,3,5dimethylbenzyl bromide
141334
1-(bromomethyl)-3,5-dimethylbenzene
CC1=CC(=CC(=C1)CBr)C
27129-86-8
C9H11Br
199.09
MFCD00013539
QXDHXCVJGBTQMK-UHFFFAOYSA-N
3,5-dimethylbenzyl bromide,1-bromomethyl-3,5-dimethylbenzene,3,5-dimethylbenzylbromide,alpha-bromomesitylene,benzene, 1-bromomethyl-3,5-dimethyl,.alpha.-bromomesitylene,5-bromomethyl-1,3-dimethylbenzene,a-bromomesitylene,pubchem9979,3,5dimethylbenzyl bromide
141334
1-(bromomethyl)-3,5-dimethylbenzene
CC1=CC(=CC(=C1)CBr)C
Packaging Glass bottle
Boiling Point 101°C to 103°C (8.0mmHg)
Color Yellow
Quantity 100g
Melting Point 36°C to 40°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 199.09
Physical Form Low Melting Crystalline Mass
Percent Purity 98%
Chemical Name or Material 3, 5-Dimethylbenzyl bromide
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger