Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylisoxazole-4-boronic Acid, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass Bottle
16114-47-9
C5H8BNO3
140.933
MFCD02677945
DIIFZCPZIRQDIJ-UHFFFAOYSA-N
3,5-dimethylisoxazole-4-boronic acid,3,5-dimethylisoxazol-4-yl boronic acid,3,5-dimethyl-4-isoxazolylboronic acid,3,5-dimethylisoxazol-4-ylboronic acid,3,5-dimethyl-1,2-oxazol-4-yl boronic acid,3,5-dimethylisoxazole-4-boronicacid,3,5-dimethyl-4-isoxazoleboronic acid,3,5-dimethylisoxazol-4-yl-4-boronic acid,3,5-dimethyl-isoxazole-4-boronic acid,dimethyl-1,2-oxazol-4-yl boronic acid
2734346
(3,5-dimethyl-1,2-oxazol-4-yl)boronic acid
B(C1=C(ON=C1C)C)(O)O
16114-47-9
C5H8BNO3
140.933
MFCD02677945
DIIFZCPZIRQDIJ-UHFFFAOYSA-N
3,5-dimethylisoxazole-4-boronic acid,3,5-dimethylisoxazol-4-yl boronic acid,3,5-dimethyl-4-isoxazolylboronic acid,3,5-dimethylisoxazol-4-ylboronic acid,3,5-dimethyl-1,2-oxazol-4-yl boronic acid,3,5-dimethylisoxazole-4-boronicacid,3,5-dimethyl-4-isoxazoleboronic acid,3,5-dimethylisoxazol-4-yl-4-boronic acid,3,5-dimethyl-isoxazole-4-boronic acid,dimethyl-1,2-oxazol-4-yl boronic acid
2734346
(3,5-dimethyl-1,2-oxazol-4-yl)boronic acid
B(C1=C(ON=C1C)C)(O)O
Quantity 250mg
Packaging Glass Bottle
Assay Percent Range 98%
Formula Weight 140.93
Chemical Name or Material 3, 5-Dimethylisoxazole-4-boronic acid