Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dimethylisoxazole-4-boronic acid pinacol ester, 97%

Click to view available options
Quantity:
5g
25g
832114-00-8
C11H18BNO3
223.079
MFCD05863910
CVLHETBAROWASE-UHFFFAOYSA-N
3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isoxazole,3,5-dimethylisoxazole-4-boronic acid pinacol ester,3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl isoxazole,3,5-dimethylisoxazole-4-boronic acid, pinacol ester,3,5-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole,3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole,3,5-dimethyl-4-isoxazolyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-3,5-dimethylisoxazol-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,pubchem18439,acmc-209prk
2758656
3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
B1(OC(C(O1)(C)C)(C)C)C2=C(ON=C2C)C
832114-00-8
C11H18BNO3
223.079
MFCD05863910
CVLHETBAROWASE-UHFFFAOYSA-N
3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isoxazole,3,5-dimethylisoxazole-4-boronic acid pinacol ester,3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl isoxazole,3,5-dimethylisoxazole-4-boronic acid, pinacol ester,3,5-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole,3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole,3,5-dimethyl-4-isoxazolyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-3,5-dimethylisoxazol-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,pubchem18439,acmc-209prk
2758656
3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
B1(OC(C(O1)(C)C)(C)C)C2=C(ON=C2C)C
Quantity 5g
Melting Point 95°C to 99°C
Color Brown to White
Formula Weight 223.08
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3,5-Dimethylisoxazole-4-boronic acid pinacol ester

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning