Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylnitrobenzene 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
99-12-7
C8H9NO2
151.17
MFCD00007269
BYFNZOKBMZKTSC-UHFFFAOYSA-N
5-nitro-m-xylene, 3,5-dimethylnitrobenzene, 3,5-dimethyl-1-nitrobenzene, benzene, 1,3-dimethyl-5-nitro, nitroxylol, m-xylene, 5-nitro, unii-5hvw2qtf0d, ccris 3121, 5hvw2qtf0d, 5-nitro-m-xylol
7426
1,3-dimethyl-5-nitrobenzene
CC1=CC(=CC(C)=C1)[N+]([O-])=O
99-12-7
C8H9NO2
151.17
MFCD00007269
BYFNZOKBMZKTSC-UHFFFAOYSA-N
5-nitro-m-xylene, 3,5-dimethylnitrobenzene, 3,5-dimethyl-1-nitrobenzene, benzene, 1,3-dimethyl-5-nitro, nitroxylol, m-xylene, 5-nitro, unii-5hvw2qtf0d, ccris 3121, 5hvw2qtf0d, 5-nitro-m-xylol
7426
1,3-dimethyl-5-nitrobenzene
CC1=CC(=CC(C)=C1)[N+]([O-])=O
Color Yellow
Boiling Point 273°C
Quantity 5g
Melting Point 73°C
UN Number 3447
Formula Weight 151.17
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3,5-Dimethylnitrobenzene

EINECSNumber : (3)-0399

TSCA : Yes