Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3,5-Dimethylphenyl isocyanate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
3,5-Dimethylphenyl isocyanate is applications include linking with oligosaccharides and cyclodextrins for chiral chromatography stationary phases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dimethylphenyl isocyanate is applications include linking with oligosaccharides and cyclodextrins for chiral chromatography stationary phases.
Solubility
Hydrolyzes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
3,5-Dimethylphenyl isocyanate is applications include linking with oligosaccharides and cyclodextrins for chiral chromatography stationary phases.
Solubility
Hydrolyzes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Density | 1.045 |
Boiling Point | 205°C |
Quantity | 1 g |
Refractive Index | 1.528 |
UN Number | UN2206 |
Beilstein | 774958 |
Solubility Information | Hydrolyzes in water. |
Formula Weight | 147.18 |
Percent Purity | 97% |
Chemical Name or Material | 3,5-Dimethylphenyl isocyanate |
Safety and Handling
Hazard Category | H227-H302+H312+H332-H315-H319-H334-H335 |
Hazard Statement | GHS H Statement H300-H330-H334-H227-H315-H319-H335 Fatal if swallowed. Fatal if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Combustible liquid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P235-P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P370+P378q-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 258-987-0 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction