Alfa Aesar™ 3,5-Dimethylphenyl isothiocyanate, 99%

Manufacturer: Alfa Aesar™ A1130222

Quantity 100g

 View more versions of this product

Catalog No. AAA1130222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 40046-30-8
    Molecular Formula C9H9NS
    Formula Weight 163.24
    MDL Number MFCD00041082
    UN Number UN2922
    Beilstein 6856812
    Chemical Name or Material 3,5-Dimethylphenyl isothiocyanate

    Specifications

    Formula Weight 163.24
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00041082
    Molecular Formula C9H9NS
    Beilstein 6856812
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H314-H318-H302-H312
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    CAS 40046-30-8
    UN Number UN2922
    Quantity 100g
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
    Chemical Name or Material 3,5-Dimethylphenyl isothiocyanate
    Assay 99%

    GHS H Statement
    H331-H314-H318-H302-H312
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit