Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylphenylboronic acid, 97%

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
172975-69-8
C8H11BO2
149.984
DJGHSJBYKIQHIK-UHFFFAOYSA-N
3,5-dimethylphenyl boronic acid,3,5-dimethylbenzeneboronic acid,3,5-dimethylphenyl boranediol,m-xylene-5-boronic acid,boronic acid, 3,5-dimethylphenyl,pubchem1833,acmc-209e5l,ksc174i6l,3.5-dimethylphenylboronic acid
2734349
(3,5-dimethylphenyl)boronic acid
B(C1=CC(=CC(=C1)C)C)(O)O
172975-69-8
C8H11BO2
149.984
DJGHSJBYKIQHIK-UHFFFAOYSA-N
3,5-dimethylphenyl boronic acid,3,5-dimethylbenzeneboronic acid,3,5-dimethylphenyl boranediol,m-xylene-5-boronic acid,boronic acid, 3,5-dimethylphenyl,pubchem1833,acmc-209e5l,ksc174i6l,3.5-dimethylphenylboronic acid
2734349
(3,5-dimethylphenyl)boronic acid
B(C1=CC(=CC(=C1)C)C)(O)O
Color White to Yellow
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 149.98
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 3, 5-Dimethylphenylboronic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning