Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylpiperidine, 96%, mixture of cis and trans, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
35794-11-7
C7H15N
113.2
MFCD00005996
IDWRJRPUIXRFRX-UHFFFAOYSA-N
piperidine, 3,5-dimethyl,3,5-lupetidine,3,5-dimethylpiperidin,3,5-dimethylpiperidine, cis + trans,pubchem7709,3,5-dimethylpiperdine,3,5 dimethylpiperidine,3,5-dimethylpiperadine,acmc-1afvq,hexahydro-3,5-lutidine
118259
3,5-dimethylpiperidine
CC1CC(CNC1)C
35794-11-7
C7H15N
113.2
MFCD00005996
IDWRJRPUIXRFRX-UHFFFAOYSA-N
piperidine, 3,5-dimethyl,3,5-lupetidine,3,5-dimethylpiperidin,3,5-dimethylpiperidine, cis + trans,pubchem7709,3,5-dimethylpiperdine,3,5 dimethylpiperidine,3,5-dimethylpiperadine,acmc-1afvq,hexahydro-3,5-lutidine
118259
3,5-dimethylpiperidine
CC1CC(CNC1)C
Boiling Point 144°C
Flash Point 32°C
Specific Gravity 0.85
Refractive Index 1.4434 to 1.4464
Density 0.8500g/mL
Water 0.5% max.
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Formula Weight 113.2
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 3, 5-Dimethylpiperidine, 96%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 252-730-6

TSCA : TSCA