Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3,5-Dimethylpiperidine, cis + trans, 97%
Click to view available options
Quantity:
25g
100g
Specifications
Refractive Index | 1.4454 |
Density | 0.83 |
Flash Point | 32°C (89°F) |
Quantity | 25g |
Boiling Point | 142°C to 146°C |
UN Number | UN1993 |
Beilstein | 102638 |
Formula Weight | 113.2 |
Percent Purity | 97% |
Chemical Name or Material | 3,5-Dimethylpiperidine, cis + trans |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 252-730-6
TSCA : Yes
Recommended Storage : Ambient temperatures