Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylpyrazole, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
67-51-6
C5H8N2
96.13
SDXAWLJRERMRKF-UHFFFAOYSA-N
3,5-dimethylpyrazole,1h-pyrazole, 3,5-dimethyl,3,5 dmp,pyrazole, 3,5-dimethyl,3,5-dwumetylopirazolu,unii-h21n865k9j,3,5-dwumetylopirazolu polish,3,5-dimethyl pyrazole,pyrazole,5-dimethyl,pubchem13256
6210
3,5-dimethyl-1H-pyrazole
CC1=CC(=NN1)C
67-51-6
C5H8N2
96.13
SDXAWLJRERMRKF-UHFFFAOYSA-N
3,5-dimethylpyrazole,1h-pyrazole, 3,5-dimethyl,3,5 dmp,pyrazole, 3,5-dimethyl,3,5-dwumetylopirazolu,unii-h21n865k9j,3,5-dwumetylopirazolu polish,3,5-dimethyl pyrazole,pyrazole,5-dimethyl,pubchem13256
6210
3,5-dimethyl-1H-pyrazole
CC1=CC(=NN1)C
Packaging Plastic bottle
Quantity 100g
Color White
Boiling Point 218.0°C
Melting Point 106.0°C to 109.0°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 23,74
Solubility Information Solubility in water: soluble
Formula Weight 96.13
Physical Form Crystalline Powder or Crystals
Percent Purity 99%
Chemical Name or Material 3, 5-Dimethylpyrazole
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.