Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-dinitroaniline 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
618-87-1
C6H5N3O4
183.123
MFCD00007263
MPBZUKLDHPOCLS-UHFFFAOYSA-N
benzenamine, 3,5-dinitro,aniline, 3,5-dinitro,unii-0ahr6k1n73,ccris 3109,3,5-dinitrophenylamine,3-5-dinitro-phenylamine,3,5-dinitroanilin,aniline,5-dinitro,acmc-209mx8,benzenamine,3,5-dinitro
12068
3,5-dinitroaniline
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])N
618-87-1
C6H5N3O4
183.123
MFCD00007263
MPBZUKLDHPOCLS-UHFFFAOYSA-N
benzenamine, 3,5-dinitro,aniline, 3,5-dinitro,unii-0ahr6k1n73,ccris 3109,3,5-dinitrophenylamine,3-5-dinitro-phenylamine,3,5-dinitroanilin,aniline,5-dinitro,acmc-209mx8,benzenamine,3,5-dinitro
12068
3,5-dinitroaniline
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])N
Color Brown-Yellow to Yellow
Melting Point 159°C to 163°C
Quantity 10g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (O2N)2C6H3NH2
Beilstein 12, 759
Formula Weight 183.12
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 3, 5-Dinitroaniline
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger