Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Dinitrobenzamide, 98%

Click to view available options
Quantity:
25g
100g
500g
121-81-3
C7H5N3O5
211.133
MFCD00007985
UUKWKUSGGZNXGA-UHFFFAOYSA-N
nitromide,tristat,unistat,benzamide, 3,5-dinitro,nitromide usan,component of tristat,nitroamide,component of unistat-3,unii-9duj3cmk8s,9duj3cmk8s
4511
3,5-dinitrobenzamide
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)N
121-81-3
C7H5N3O5
211.133
MFCD00007985
UUKWKUSGGZNXGA-UHFFFAOYSA-N
nitromide,tristat,unistat,benzamide, 3,5-dinitro,nitromide usan,component of tristat,nitroamide,component of unistat-3,unii-9duj3cmk8s,9duj3cmk8s
4511
3,5-dinitrobenzamide
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)N
Quantity 500g
Melting Point 181°C to 183°C
Merck Index 14,6612
Formula Weight 211.13
Percent Purity 98%
Chemical Name or Material 3,5-Dinitrobenzamide

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger