Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3,5-Dinitrobenzotrifluoride, 98%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Flash Point | >110°C (230°F) |
Melting Point | 50°C to 52°C |
Quantity | 25g |
Beilstein | 2058079 |
Formula Weight | 236.11 |
Percent Purity | 98% |
Chemical Name or Material | 3,5-Dinitrobenzotrifluoride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 206-935-2
TSCA : No
Recommended Storage : Ambient temperatures