Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Lutidine 98+%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
591-22-0
C7H9N
107.15
MFCD00006404
HWWYDZCSSYKIAD-UHFFFAOYSA-N
3,5-lutidine,pyridine, 3,5-dimethyl,unii-6q4ypz045v,3,5-dimethyl pyridine,3,5 lutidine,3,5-lutdcine,3,5-dimethylpyridin,3,5-dimethyl-pyridine,acmc-1axev,dsstox_cid_30825
11565
3,5-dimethylpyridine
CC1=CC(=CN=C1)C
591-22-0
C7H9N
107.15
MFCD00006404
HWWYDZCSSYKIAD-UHFFFAOYSA-N
3,5-lutidine,pyridine, 3,5-dimethyl,unii-6q4ypz045v,3,5-dimethyl pyridine,3,5 lutidine,3,5-lutdcine,3,5-dimethylpyridin,3,5-dimethyl-pyridine,acmc-1axev,dsstox_cid_30825
11565
3,5-dimethylpyridine
CC1=CC(=CN=C1)C
Melting Point -9.0°C
Quantity 100mL
Flash Point 53°C
Refractive Index 1.5040 to 1.5060
Packaging Glass bottle
Water 0.2% max. (K.F.)
Specific Gravity 0.93
Infrared Spectrum Authentic
Density 0.9300g/mL
Color Yellow
Boiling Point 169.0°C to 170.0°C
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Beilstein 20, 246
Solubility Information Solubility in water: 33g/L (20°C). Other solubilities: soluble in ethanol and ether
Formula Weight 107.15
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material 3, 5-Lutidine, 98+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-708-6

TSCA : TSCA