Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(5-Methyl-2-furyl)butyraldehyde, 99%

Click to view available options
Quantity:
5g
25g
100g
31704-80-0
C9H12O2
152.193
MFCD00209519
DFSVNSCDOZSUCT-UHFFFAOYSA-N
3-5-methyl-2-furyl butyraldehyde,3-5-methylfuran-2-yl butanal,3-5-methyl-2-furyl butanal,3-5-methyl-2-furanyl butanal,fema no. 3307,beta5-dimethyl-2-furanpropanal,2-furanpropanal, beta5-dimethyl,2-furanpropanal, beta,5-dimethyl,beta5-dimethyl-2-furanpropionaldehyde,2-furanpropanal, .beta.,5-dimethyl
3578033
3-(5-methylfuran-2-yl)butanal
CC1=CC=C(O1)C(C)CC=O
31704-80-0
C9H12O2
152.193
MFCD00209519
DFSVNSCDOZSUCT-UHFFFAOYSA-N
3-5-methyl-2-furyl butyraldehyde,3-5-methylfuran-2-yl butanal,3-5-methyl-2-furyl butanal,3-5-methyl-2-furanyl butanal,fema no. 3307,beta5-dimethyl-2-furanpropanal,2-furanpropanal, beta5-dimethyl,2-furanpropanal, beta,5-dimethyl,beta5-dimethyl-2-furanpropionaldehyde,2-furanpropanal, .beta.,5-dimethyl
3578033
3-(5-methylfuran-2-yl)butanal
CC1=CC=C(O1)C(C)CC=O
Quantity 100g
Boiling Point 93°C to 94°C (14mmHg)
Flash Point 58°C (136°F)
UN Number UN1989
Formula Weight 152.19
Percent Purity 99%
Chemical Name or Material 3-(5-Methyl-2-furyl)butyraldehyde

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.

TSCA : TSCA