Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Pyridinedicarboxylic acid, 98%, ACROS Organics™

missing translation for 'orderingAttributeHoverText'
Quantity:
10g
50g
Packaging:
Glass bottle
499-81-0
C7H5NO4
167.12
MPFLRYZEEAQMLQ-UHFFFAOYSA-N
3,5-pyridinedicarboxylic acid,dinicotinic acid,5-carboxynicotinic acid,3,5-pyridinedicarboxylicacid,3,5-pyridindicarbons,3,5-dipyridinecarboxylic acid,pubchem8057,acmc-209kjb,5-carboxy nicotinic acid,pyridine carboxylate, 6f
10366
CHEBI:46875
pyridine-3,5-dicarboxylic acid
C1=C(C=NC=C1C(=O)O)C(=O)O
499-81-0
C7H5NO4
167.12
MPFLRYZEEAQMLQ-UHFFFAOYSA-N
3,5-pyridinedicarboxylic acid,dinicotinic acid,5-carboxynicotinic acid,3,5-pyridinedicarboxylicacid,3,5-pyridindicarbons,3,5-dipyridinecarboxylic acid,pubchem8057,acmc-209kjb,5-carboxy nicotinic acid,pyridine carboxylate, 6f
10366
CHEBI:46875
pyridine-3,5-dicarboxylic acid
C1=C(C=NC=C1C(=O)O)C(=O)O
Packaging Glass bottle
Water 1% Max. (K.F.)
Color White
Melting Point >300.0°C
Quantity 50g
Infrared Spectrum Authentic
Assay Percent Range 98%
Beilstein 22,16
Solubility Information Solubility in water: insoluble
Formula Weight 167.12
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 3, 5-Pyridinedicarboxylic acid
Show More Show Less