Learn More
3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane, 41% solution in aromatic free mi, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 349940050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiatorsSpecifications
3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane | |
41% Solution in Aromatic Free Min. Spirit | |
40.0, 58.0 | |
42.0, 60.0 | |
MFCD02685709 | |
unii-j3mv6azn38, 3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexoxonane, 1,2,4,5,7,8-hexoxonane, 3,6,9-triethyl-3,6,9-trimethyl, j3mv6azn38, 3,6,9-triethyl-3,6,9-trimethyl-1,4,7-triperoxynonane, unii-j3mv6azn38 component kvwlloiegklbpa-aqhstmncsa-n, unii-j3mv6azn38 component kvwlloiegklbpa-zsbigdgjsa-n, 3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexaoxacyclononane, 1,2,4,5,7,8-hexaoxacyclononane, 3,6,9-trimethyl, 3,6,9-triethyl, 3,6,9-triethyl-3,6,9-trimethyl-1,4,7-triperoxynonane sol.in arom.free min.spirit | |
KVWLLOIEGKLBPA-UHFFFAOYSA-N | |
3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexaoxonane | |
2734078 | |
7.3 to 7.6% (total active oxygen) | |
0.875 | |
5 g | |
Solution |
Undesignated | |
24748-23-0, 78-93-3 | |
<10.0°C | |
C12H24O6 | |
0.8750g/mL | |
Solubility in water: immiscible. Other solubilities: miscible with most organic solvents | |
CCC1(OOC(OOC(OO1)(C)CC)(C)CC)C | |
264.318 | |
264.32 | |
Plastic bottle | |
5 mPa.s (20°C) | |
74°C |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Causes skin irritation.
May be fatal if swallowed and enters airways.
Heating may cause a fire or explosion.
Flammable liquid and vapor.
GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
TSCA : TSCA
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only