Learn More
3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane, 41% solution in aromatic free mi, ACROS Organics™
Description
Specifications
CAS Min % | 40.0,58.0 |
CAS Max % | 42.0,60.0 |
Density | 0.8750g/mL |
Formula Weight | 264.32 |
Physical Form | Solution |
Percent Purity | 7.3 to 7.6% (total active oxygen) |
Packaging | Plastic bottle |
Solubility | Solubility in water: immiscible. Other solubilities: miscible with most organic solvents |
Specific Gravity | 0.875 |
Viscosity | 5 mPa.s (20°C) |
Show More |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Causes skin irritation.
May be fatal if swallowed and enters airways.
Heating may cause a fire or explosion.
Flammable liquid and vapor.
GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
TSCA : TSCA