Learn More
Aat BioquestSupplier Diversity Partner 3,6-Dichlorobenzene-1,2,4-tricarboxylic anhydride [3,6-Dichlorotrimellitic anhydride]  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 4021
3,6-Dichlorotrimellitic anhydride is the key precursor that is used for preparing a variety of dichlorinated fluoresceins and rhodamines such as TET and HEX.
Catalog No. 50-195-3231