Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester, 98%

Click to view available options
Quantity:
250mg
1g
5g
287944-16-5
C11H19BO3
210.08
MFCD11052631
DOSGEBYQRMBTGS-UHFFFAOYSA-N
3,6-dihydro-2h-pyran-4-boronic acid pinacol ester,2-3,6-dihydro-2h-pyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-pyran,4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyran,2h-pyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyran,3,6-dihydro-2h-pyran-4-boronic acid, pinacol ester,3,6-dihydro-2h-pyran-4-boronicacidpinacolester,3,6-dihydro-2h-pyran-4-boronic acid pinacolester,3,6 dihydro-2h-pyran-4-boronic acid pinacol ester
11218053
2-(3,6-dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CCOCC2
287944-16-5
C11H19BO3
210.08
MFCD11052631
DOSGEBYQRMBTGS-UHFFFAOYSA-N
3,6-dihydro-2h-pyran-4-boronic acid pinacol ester,2-3,6-dihydro-2h-pyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-pyran,4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyran,2h-pyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyran,3,6-dihydro-2h-pyran-4-boronic acid, pinacol ester,3,6-dihydro-2h-pyran-4-boronicacidpinacolester,3,6-dihydro-2h-pyran-4-boronic acid pinacolester,3,6 dihydro-2h-pyran-4-boronic acid pinacol ester
11218053
2-(3,6-dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CCOCC2
Applications
It plays an important role as intermediate for organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
Quantity 250mg
Sensitivity Moisture Sensitive
Melting Point 63°C to 67°C
Color White
Beilstein 8548328
Formula Weight 210.08
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C