Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,6-Dihydrothiopyran-4-boronic acid pinacol ester, 98%

Click to view available options
Quantity:
250mg
1g
5g
862129-81-5
C11H19BO2S
226.141
MFCD10700154
QZVRTORVCKGPPY-UHFFFAOYSA-N
2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,3,6-dihydro-2h-thiopyran-4-ylboronic acid pinacol ester,2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane,2h-thiopyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,6-dihydro-2h-thiopyran-4-yl-4,3,6-dihydrothiopyran-4-boronic acid pinacol ester,3,6-dihydro-2h-thiopyran-4-ylboronicacidpinacolester,3,6-dihydro-2h-thiopyran-4-yl boronic acid pinacol ester,2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5,-tetramethyl-1,3,2-dioxaborolane
46739735
2-(3,6-dihydro-2H-thiopyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CCSCC2
862129-81-5
C11H19BO2S
226.141
MFCD10700154
QZVRTORVCKGPPY-UHFFFAOYSA-N
2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,3,6-dihydro-2h-thiopyran-4-ylboronic acid pinacol ester,2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane,2h-thiopyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,6-dihydro-2h-thiopyran-4-yl-4,3,6-dihydrothiopyran-4-boronic acid pinacol ester,3,6-dihydro-2h-thiopyran-4-ylboronicacidpinacolester,3,6-dihydro-2h-thiopyran-4-yl boronic acid pinacol ester,2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5,-tetramethyl-1,3,2-dioxaborolane
46739735
2-(3,6-dihydro-2H-thiopyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CCSCC2
Quantity 1g
Beilstein 11090000
Formula Weight 226.14
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 3,6-Dihydrothiopyran-4-boronic acid pinacol ester

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning