Learn More
Alfa Aesar™ 3,7-Dichloro-6-methoxybenzo[b]thiophene-2-carbonyl chloride
Manufacturer:  Alfa Aesar™ H50424-MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH50424MD
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
34576-80-2
295.57
UN3261
C10H5Cl3O2S
MFCD02090812
3,7-Dichloro-6-methoxybenzo[b]thiophene-2-carbonyl chloride
Specifications
250mg
C10H5Cl3O2S
MFCD02090812
3,7-Dichloro-6-methoxybenzo[b]thiophene-2-carbonyl chloride
34576-80-2
295.57
UN3261
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.