Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,8-Diamino-6-phenylphenanthridine, 98%

Click to view available options
Quantity:
1g
5g
25g
52009-64-0
C19H15N3
285.35
MFCD00075152
CPNAVTYCORRLMH-UHFFFAOYSA-N
3,8-diamino-6-phenylphenanthridine,6-phenylphenathridine-3,8-diamine,3,8-dapp,3,8-diamino-6-phenyl-phenanthridine,acmc-1aycj,6-phenyl-3,8-phenanthridinediamine,3,8-diamino-6-phenyl phenanthridine,3,8-phenanthridinediamine,6-phenyl,3,8-diamino-6-phenylphenanthridine6-phenyl-3,8-phenanthridinediamine,
104052
6-phenylphenanthridine-3,8-diamine
C1=CC=C(C=C1)C2=C3C=C(C=CC3=C4C=CC(=CC4=N2)N)N
52009-64-0
C19H15N3
285.35
MFCD00075152
CPNAVTYCORRLMH-UHFFFAOYSA-N
3,8-diamino-6-phenylphenanthridine,6-phenylphenathridine-3,8-diamine,3,8-dapp,3,8-diamino-6-phenyl-phenanthridine,acmc-1aycj,6-phenyl-3,8-phenanthridinediamine,3,8-diamino-6-phenyl phenanthridine,3,8-phenanthridinediamine,6-phenyl,3,8-diamino-6-phenylphenanthridine6-phenyl-3,8-phenanthridinediamine,
104052
6-phenylphenanthridine-3,8-diamine
C1=CC=C(C=C1)C2=C3C=C(C=CC3=C4C=CC(=CC4=N2)N)N
Applications
3,8-Diamino-6-phenylphenanthridine is used in the synthesis of rigid polyamides. It has anti-tumor and anti-viral properties.

Solubility
Soluble in chloroform, dichloromethane and methanol.

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Quantity 1g
Melting Point 196°C to 198°C
Solubility Information Soluble in chloroform,dichloromethane and methanol.
Formula Weight 285.35
Percent Purity 98%
Chemical Name or Material 3,8-Diamino-6-phenylphenanthridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 257-602-3

TSCA : No

Recommended Storage : Ambient temperatures