Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,8-Diamino-6-phenylphenanthridine, 98%

Click to view available options
Quantity:
1g
5g
25g
52009-64-0
C19H15N3
285.35
MFCD00075152
CPNAVTYCORRLMH-UHFFFAOYSA-N
3,8-diamino-6-phenylphenanthridine,6-phenylphenathridine-3,8-diamine,3,8-dapp,3,8-diamino-6-phenyl-phenanthridine,acmc-1aycj,6-phenyl-3,8-phenanthridinediamine,3,8-diamino-6-phenyl phenanthridine,3,8-phenanthridinediamine,6-phenyl,3,8-diamino-6-phenylphenanthridine6-phenyl-3,8-phenanthridinediamine,
104052
6-phenylphenanthridine-3,8-diamine
C1=CC=C(C=C1)C2=C3C=C(C=CC3=C4C=CC(=CC4=N2)N)N
52009-64-0
C19H15N3
285.35
MFCD00075152
CPNAVTYCORRLMH-UHFFFAOYSA-N
3,8-diamino-6-phenylphenanthridine,6-phenylphenathridine-3,8-diamine,3,8-dapp,3,8-diamino-6-phenyl-phenanthridine,acmc-1aycj,6-phenyl-3,8-phenanthridinediamine,3,8-diamino-6-phenyl phenanthridine,3,8-phenanthridinediamine,6-phenyl,3,8-diamino-6-phenylphenanthridine6-phenyl-3,8-phenanthridinediamine,
104052
6-phenylphenanthridine-3,8-diamine
C1=CC=C(C=C1)C2=C3C=C(C=CC3=C4C=CC(=CC4=N2)N)N
Applications
3,8-Diamino-6-phenylphenanthridine is used in the synthesis of rigid polyamides. It has anti-tumor and anti-viral properties.

Solubility
Soluble in chloroform, dichloromethane and methanol.

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Melting Point 196°C to 198°C
Quantity 25g
Formula Weight 285.35
Percent Purity 98%
Chemical Name or Material 3,8-Diamino-6-phenylphenanthridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning