Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(9-Anthryl)acrolein 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
38982-12-6
C17H12O
232.282
MFCD09751223
NLHDGEXLFCDTHQ-BJMVGYQFSA-N
3-(9-Anthryl)propenal
6365417
(E)-3-anthracen-9-ylprop-2-enal
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2C=CC=O
38982-12-6
C17H12O
232.282
MFCD09751223
NLHDGEXLFCDTHQ-BJMVGYQFSA-N
3-(9-Anthryl)propenal
6365417
(E)-3-anthracen-9-ylprop-2-enal
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2C=CC=O
Melting Point 172°C
Color Undesignated
Quantity 5g
Formula Weight 232.28
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3-(9-Anthryl)acrolein